Learn More
Quality Biological Inc LB Agar with 100 g/mL Ampicillin, CS 100 X 40 mL
SDP
Supplier:  Quality Biological Inc 340108211
Encompass_Preferred
LB Agar Plates are used for the growth and maintenance of E. coli strains used in molecular microbiology procedures. ¿ Applications: Nutritionally rich media designed for growth of pure cultures of recombinant strains. LB Agar plates are suitable for propagation of E. Coli and other bacteriological stocks. ¿ Storage Conditions: 2¿C to 8¿C ¿ Shipping Condition: ambient temperatureAutoclave Sterilized ¿ UNSPSC: 41106200 ¿ ¿ Max Shelf Life: NA
Catalog No. 50-751-7581