Learn More
Grainger LB Swivel Valve Stem 2-1/2
Catalog No. 502753019
Encompass_Preferred
The Grainger 141D41 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long and weighs 0.19 lbs. The 141D41 socket adapter is ideal for automotive repair shops, fleet maintenance, and other heavy duty applications requiring high torque ratcheting and driving.
Catalog No. 50-275-3019 Supplier Grainger Supplier No. 141D41
The Grainger 141D41 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long and weighs 0.19 lbs. The 141D41 socket adapter is ideal for automotive repair shops, fleet maintenance, and other heavy duty applications requiring high torque ratcheting and driving.