Learn More
Teknova LB Top Agar. 100mL, Sterile.
SDP
Catalog No. 502388002
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 0.8% Agar
Catalog No. 50-238-8002 Supplier Teknova Supplier No. L5580
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 0.8% Agar