Learn More
Teknova LB Top Agar. 454 grams.
SDP
Catalog No. 502388178
Encompass
1.00% Tryptone, 0.50% Yeast Extract,1.00% Sodium Chloride, 0.75% Agar
Catalog No. 50-238-8178 Supplier Teknova Supplier No. L9155
1.00% Tryptone, 0.50% Yeast Extract,1.00% Sodium Chloride, 0.75% Agar