Learn More
Apexbio Technology LLC LB42708, 10mg. Cas: 226929-39-1 MFCD: MFCD20926350. Selective FTase inhibitor
SDP
Supplier:  Apexbio Technology LLC B370310
Encompass_Preferred
LB42708 is a selective inhibitor of farnesyltransferase (FTase) with IC50 value of 0.8nM [1].LB42708 is a nonpeptide pyrrole-based FTase inhibitor with high potency and selectivity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5351