Learn More
Apexbio Technology LLC LB42708 226929-39-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063085
Encompass_Preferred
LB42708 (CAS 226929-39-1) is a selective small-molecule inhibitor of farnesyltransferase (FTase) belonging to a non-peptidic pyrrole-based class It blocks FTase-mediated farnesylation of H-Ras N-Ras and K-Ras4B demonstrated by low nanomolar IC50 values (0 8 nM 1 2 nM and 2 nM respectively) LB42708 shows minimal inhibitory effect on geranylgeranyltransferase I In vitro studies indicate inhibition of Ras-driven signaling pathways decreasing Ras activation and subsequently suppressing VEGF-stimulated endothelial proliferation and migration Researchers use LB42708 in oncology and inflammation studies exemplified by reduced tumor growth in Ras-mutant HCT116 and wild-type Caco-2 xenograft models
Catalog No. 50-000-63085 Supplier Apexbio Technology LLC Supplier No. B37035.1
LB42708 (CAS 226929-39-1) is a selective small-molecule inhibitor of farnesyltransferase (FTase) belonging to a non-peptidic pyrrole-based class It blocks FTase-mediated farnesylation of H-Ras N-Ras and K-Ras4B demonstrated by low nanomolar IC50 values (0 8 nM 1 2 nM and 2 nM respectively) LB42708 shows minimal inhibitory effect on geranylgeranyltransferase I In vitro studies indicate inhibition of Ras-driven signaling pathways decreasing Ras activation and subsequently suppressing VEGF-stimulated endothelial proliferation and migration Researchers use LB42708 in oncology and inflammation studies exemplified by reduced tumor growth in Ras-mutant HCT116 and wild-type Caco-2 xenograft models