Learn More
INTACT GENOMICS INC LBA4404 Agrobacterium Chemically Competent Cells 10x50µl
Catalog No. 5000083054
Encompass
Intact Genomics LBA4404 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies which is ideal for applications such as cDNA or gDNA library construction. The LBA4404 strain is useful for transgenic operations of tomatoes, tobacco and other plants. LBA4404 contains a rifampicin resistance gene (rif). LBA4404 strain also contains an octopine-type Ti plasmid pAL4404 without self-transport function, which contains the vir gene. Product Includes and Storage:LBA4404 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-83054 Supplier INTACT GENOMICS INC Supplier No. 108510
Intact Genomics LBA4404 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies which is ideal for applications such as cDNA or gDNA library construction. The LBA4404 strain is useful for transgenic operations of tomatoes, tobacco and other plants. LBA4404 contains a rifampicin resistance gene (rif). LBA4404 strain also contains an octopine-type Ti plasmid pAL4404 without self-transport function, which contains the vir gene. Product Includes and Storage:LBA4404 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC