Learn More
Gold Biotechnology Inc LBA4404 Agro Electro Cells
SDP
Catalog No. 5000562330
Encompass_Preferred
GoldBio s LBA4404 Electrocompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our LBA4404 strain harbor a rifampicin resistance (rif) gene Furthermore LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function containing the vir genes Our LBA4404 strain can be used in genetic transformation of tomato tobacco and other plants
Catalog No. 50-005-62330 Supplier Gold Biotechnology Inc Supplier No. CC22015X50
GoldBio s LBA4404 Electrocompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our LBA4404 strain harbor a rifampicin resistance (rif) gene Furthermore LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function containing the vir genes Our LBA4404 strain can be used in genetic transformation of tomato tobacco and other plants