Learn More
INTACT GENOMICS INC Methionine Auxotrophic LBA4404 Agrobacterium Chemically Competent Cells 5x50µl
Supplier:  INTACT GENOMICS INC 107605
Encompass
Intact Genomics (ig) Methionine-Auxotrophic LBA4404Met Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless free methionine is added to minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Methionine- Auxotrophic LBA4404Met Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies. The LBA4404Met strain is useful for transgenic operations of tomatoes, tobacco and other plants. LBA4404Met contains a rifampicin resistance gene (rif). LBA4404Met strain also contains an octopine-type Ti plasmid pAL4404Met without self-transport function, which contains the vir gene.Benefits: Methionine Auxotrophic Enables development of more efficient transformation systems Reduced bacterial overgrowth during co-cultivation Decreased need for antibiotics
Catalog No. 50-000-83053