Learn More
Gold Biotechnology Inc LBA4404 Chemically Competent Agrobacterium 5x50µL
SDP
Catalog No. NC2942841
Encompass_Preferred
GoldBio’s LBA4404 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our LBA4404 strain harbors a rifampicin resistance (rif) gene. Furthermore, LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function, containing the vir genes. Our LBA4404 strain can be used in genetic transformation of tomato, tobacco and other plants.
Catalog No. NC2942841 Supplier Gold Biotechnology Inc Supplier No. CC1075X50
GoldBio’s LBA4404 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our LBA4404 strain harbors a rifampicin resistance (rif) gene. Furthermore, LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function, containing the vir genes. Our LBA4404 strain can be used in genetic transformation of tomato, tobacco and other plants.