Learn More
Gold Biotechnology Inc LBA4404 pSuperAgro v4 Comp
SDP
Catalog No. 5000562207
Encompass_Preferred
GoldBio s LBA4404 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our LBA4404 strain harbors a rifampicin resistance (rif) gene Furthermore LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function containing the vir genes Our LBA4404 strain can be used in genetic transformation of tomato tobacco and other plants
Catalog No. 50-005-62207 Supplier Gold Biotechnology Inc Supplier No. CC54415X50
GoldBio s LBA4404 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our LBA4404 strain harbors a rifampicin resistance (rif) gene Furthermore LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function containing the vir genes Our LBA4404 strain can be used in genetic transformation of tomato tobacco and other plants