Learn More
Cayman Chemical LBiopterin Fatty Acids and Der
SDP
Catalog No. 502889771
Encompass_Preferred
An intermediate for tetrahydro-L-biopterin; oxidized form of BH4, a NOS cofactor; toxic to human melanocytes in culture (IC50 = 0.2 µM after 48 hrs); rarely found under physiological conditions except in the epidermis of patients with Vitiligo
Catalog No. 50-288-9771 Supplier Cayman Chemical Supplier No. 1000766250 MG
An intermediate for tetrahydro-L-biopterin; oxidized form of BH4, a NOS cofactor; toxic to human melanocytes in culture (IC50 = 0.2 µM after 48 hrs); rarely found under physiological conditions except in the epidermis of patients with Vitiligo