Learn More
Cayman Chemical LBiopterin Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 1000766210 MG
Encompass_Preferred
An intermediate for tetrahydro-L-biopterin; oxidized form of BH4, a NOS cofactor; toxic to human melanocytes in culture (IC50 = 0.2 µM after 48 hrs); rarely found under physiological conditions except in the epidermis of patients with Vitiligo
Catalog No. 50-288-8169