Learn More
FUJIFILM Biosciences LBIS Human Apo B-48 Control Se
Catalog No. 5000450380
Encompass_Preferred
This product is a precision control set for the LBIS Human Apo B-48 ELISA Kit 298-88901 The set includes Low control and High control which can be used as is after thawing Please use this set for precision control in each measurement Includes both high and low control levels L and H For research use only RUO
Catalog No. 50-004-50380 Supplier FUJIFILM Biosciences Supplier No. 29489003
This product is a precision control set for the LBIS Human Apo B-48 ELISA Kit 298-88901 The set includes Low control and High control which can be used as is after thawing Please use this set for precision control in each measurement Includes both high and low control levels L and H For research use only RUO