Learn More
Saf T Pak Inc STP-806 - Cargo Aircraft Only Label, 5 x 5in.
SDP
Supplier:  Saf T Pak Inc STP806
Encompass_Preferred
Cargo Aircraft Only Label, Use when shipping Infectious substances in quantities more than 50g or 50ml, Dimensions: 5 x 5 Inches / 125mm x 125mm, 120/Case
Catalog No. NC0076933