Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC AccuformNMC HazMat Label, 1 x 3 in, Removable Adhesive, 500/Roll

Catalog No. 501909721
Encompass

This hazardous material label is designed for clear identification of chemicals, owners, and dates. It features a removable adhesive, is made from durable paper material, and comes in a convenient roll for easy dispensing.

  • Removable adhesion for flexible application
  • Pressure-sensitive paper material
  • Hazardous Material message type
  • Legend includes fields for chemical, owner, and date
  • English language
  • Overall height of 1 in.
  • Graphic and message with diamond boxes
  • Multi-color (Red, White, Blue, Yellow, Black)
  • Length of 3 in.
  • Graffiti-proof, chemical-resistant, and waterproof properties
  • Roll package type
  • Supplied as a 500 piece roll

Catalog No. 50-190-9721 Supplier MSC Supplier No. 63376693
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This hazardous material label is designed for clear identification of chemicals, owners, and dates. It features a removable adhesive, is made from durable paper material, and comes in a convenient roll for easy dispensing.

  • Removable adhesion for flexible application
  • Pressure-sensitive paper material
  • Hazardous Material message type
  • Legend includes fields for chemical, owner, and date
  • English language
  • Overall height of 1 in.
  • Graphic and message with diamond boxes
  • Multi-color (Red, White, Blue, Yellow, Black)
  • Length of 3 in.
  • Graffiti-proof, chemical-resistant, and waterproof properties
  • Roll package type
  • Supplied as a 500 piece roll

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.