Learn More
GA International DYMO LabelWriter™ 5-Series Cryogenic Labels for Frozen Surfaces 0.5" / 12.7mm Circle White 2000 labels/roll
Catalog No. NC3454656
Encompass_Preferred
CryoSTUCK labels for frozen surfaces are compatible with DYMO LabelWriter 5-Series printers for easy and reliable printing. These durable, solvent-resistant cryogenic labels are designed for direct application to frozen surfaces as low as (-80°C/-112°F), including on vials and tubes stored in ultra-low temperature freezers. These labels can also be applied to room temperature vials prior to storage in ultra-low temperature freezers, ensuring secure adhesion and durability throughout the entire freezing process. They offer a reliable way to label frozen containers without needing to thaw samples first, ensuring the integrity of valuable temperature-sensitive specimens.The labels feature a permanent adhesive that remains secure under extreme conditions, including prolonged liquid nitrogen storage (-196°C/-321°F). Once applied, the labels ensure long-term preservation and stability, ideal for research institutes, clinical laboratories, biobanks, and biotechnology companies.Additional
Catalog No. NC3454656 Supplier GA International Supplier No. LWCS510WH
CryoSTUCK labels for frozen surfaces are compatible with DYMO LabelWriter 5-Series printers for easy and reliable printing. These durable, solvent-resistant cryogenic labels are designed for direct application to frozen surfaces as low as (-80°C/-112°F), including on vials and tubes stored in ultra-low temperature freezers. These labels can also be applied to room temperature vials prior to storage in ultra-low temperature freezers, ensuring secure adhesion and durability throughout the entire freezing process. They offer a reliable way to label frozen containers without needing to thaw samples first, ensuring the integrity of valuable temperature-sensitive specimens.The labels feature a permanent adhesive that remains secure under extreme conditions, including prolonged liquid nitrogen storage (-196°C/-321°F). Once applied, the labels ensure long-term preservation and stability, ideal for research institutes, clinical laboratories, biobanks, and biotechnology companies.Additional