Learn More
GA International DYMO LabelWriter™ 5-Series Cryogenic Labels for Frozen Surfaces 2" x 1" / 50.8mm x 25.4mm Rectangle White 1000 labels/roll
Supplier:  GA International LWCS501WH
Encompass_Preferred
CryoSTUCK labels for frozen surfaces are compatible with DYMO LabelWriter 5-Series printers for easy and reliable printing. These durable, solvent-resistant cryogenic labels are designed for direct application to frozen surfaces as low as (-80°C/-112°F), including on vials and tubes stored in ultra-low temperature freezers. These labels can also be applied to room temperature vials prior to storage in ultra-low temperature freezers, ensuring secure adhesion and durability throughout the entire freezing process. They offer a reliable way to label frozen containers without needing to thaw samples first, ensuring the integrity of valuable temperature-sensitive specimens.The labels feature a permanent adhesive that remains secure under extreme conditions, including prolonged liquid nitrogen storage (-196°C/-321°F). Once applied, the labels ensure long-term preservation and stability, ideal for research institutes, clinical laboratories, biobanks, and biotechnology companies.Additional
Catalog No. NC3454655