Learn More
Labelmaster Inc LBL NON RCRA REGULATED 100/RL
SDP
Catalog No. NC1956498
Encompass_Preferred
Play it safe when shipping non-RCRA regulated waste with Labelmaster's Non-RCRA Regulated Waste Labels for 12mm UN/NA numbers. These labels are designed to accommodate up to a 12mm UN/NA number to meet the 49 CFR 172.301 marking requirements that became effective January 1, 2017. Using your PC and printer, their ruled lines allow you to fill in the information necessary for marking a container for transport in compliance with local EPA and DOT regulations. Constructed of thermal PVC-free film, these flexible labels feature automotive-grade ink pigments and specially formulated adhesives. They also pass the 90-day IMO seawater immersion test. They measure 6 x 6 in. and are sold in rolls of 100. LABELMASTER INC mfg# BWMTL12
Catalog No. NC1956498 Supplier Labelmaster Inc Supplier No. BWMTL12
Play it safe when shipping non-RCRA regulated waste with Labelmaster's Non-RCRA Regulated Waste Labels for 12mm UN/NA numbers. These labels are designed to accommodate up to a 12mm UN/NA number to meet the 49 CFR 172.301 marking requirements that became effective January 1, 2017. Using your PC and printer, their ruled lines allow you to fill in the information necessary for marking a container for transport in compliance with local EPA and DOT regulations. Constructed of thermal PVC-free film, these flexible labels feature automotive-grade ink pigments and specially formulated adhesives. They also pass the 90-day IMO seawater immersion test. They measure 6 x 6 in. and are sold in rolls of 100. LABELMASTER INC mfg# BWMTL12