Learn More
Midwest Scientific 3/4'' Label Tape, Assorted colors, 500"
SDP
Catalog No. NC1445664
Encompass
Color coding tape is easy to write on and removes easily without leaving any sticky residue behind. The tape is strong, durable, oil resistant, water proof, and acid resistant. It can withstand temperatures from -23° to +121° Celsius (10° to +250° Fahrenheit). Colored Tape can be written on with pen, pencil, or permanent marker. Tape will stick to most surfaces including glass, plastic, and metal.
Catalog No. NC1445664 Supplier Midwest Scientific Supplier No. MIDSCIT2427
Color coding tape is easy to write on and removes easily without leaving any sticky residue behind. The tape is strong, durable, oil resistant, water proof, and acid resistant. It can withstand temperatures from -23° to +121° Celsius (10° to +250° Fahrenheit). Colored Tape can be written on with pen, pencil, or permanent marker. Tape will stick to most surfaces including glass, plastic, and metal.