Learn More
Research Products International Corp Tough-Tags, Laser Sheets, 0.94 x 0.5 Inch Labels for 0.5-0.65ml Tubes, RNBW Colors, 2975/pk
SDP
Catalog No. 502229565
Encompass
Tough-Tag, Laser Sheets, are pre-cut, peel-off labels that fit micro-tubes and a variety of other containers in research laboratories. Labels are manufactured from chemically inert polyester with strong adhesion properties. Tough-Tags adhere to all plastics and withstand steam autoclaving, boiling water baths and freezer temperatures, organic solvents and caustic agents. Supplied in Rainbow pack of colors.
Tough-Tags are made of a special smear resistant material and will accept any marking instrument.
Catalog No. 50-222-9565 Supplier Research Products International Corp Supplier No. 247193RAINBOW
Tough-Tag, Laser Sheets, are pre-cut, peel-off labels that fit micro-tubes and a variety of other containers in research laboratories. Labels are manufactured from chemically inert polyester with strong adhesion properties. Tough-Tags adhere to all plastics and withstand steam autoclaving, boiling water baths and freezer temperatures, organic solvents and caustic agents. Supplied in Rainbow pack of colors.
Tough-Tags are made of a special smear resistant material and will accept any marking instrument.