Learn More
Aviva Systems Biology Corp LBP Antibody - middle region (ARP41545_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41545_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LBP. Alternate names: MGC22233, BPIFD2ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57896