Learn More
Aviva Systems Biology Corp LBP Antibody - C-terminal region (ARP41546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LBP. Alternate names: MGC22233, BPIFD2ApplicationWB, IHC. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-57897