Learn More
Medchemexpress LLC Lipopolysaccharide-binding protein (human, HEK293, His) | 100UG
SDP
Catalog No. 5000174352
Encompass_Preferred
Recombinant human lipopolysaccharide-binding protein produced in HEK293 cells with a C-terminal His tag. LBP binds the lipid A moiety of bacterial LPS and facilitates CD14 interaction, making it suitable for studies of innate immune signaling, LPS-binding assays, and use as a positive control in immunoassays.
  • Mammalian expression in HEK293 for native glycosylation.
  • C-terminal His tag for purification and detection.
  • Full-length sequence corresponds to residues A26-V481 (456 aa).
  • Apparent molecular weight ~55-75 kDa due to glycosylation.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Datasheet guidance: avoid reconstituting below 100 μg/mL.
Catalog No. 50-001-74352 Supplier Medchemexpress LLC Supplier No. HYP70313100UG
Recombinant human lipopolysaccharide-binding protein produced in HEK293 cells with a C-terminal His tag. LBP binds the lipid A moiety of bacterial LPS and facilitates CD14 interaction, making it suitable for studies of innate immune signaling, LPS-binding assays, and use as a positive control in immunoassays.
  • Mammalian expression in HEK293 for native glycosylation.
  • C-terminal His tag for purification and detection.
  • Full-length sequence corresponds to residues A26-V481 (456 aa).
  • Apparent molecular weight ~55-75 kDa due to glycosylation.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Datasheet guidance: avoid reconstituting below 100 μg/mL.