Learn More
Hycult Biotech Inc LBP MOUSE ELISA KIT 2X96 DET
SDP
Supplier:  Hycult Biotech Inc HK20502
Encompass
The mouse LBP ELISA has been developed for the quantitative measurement of natural mouse LBP in cell culture medium, plasma or serum.
Catalog No. NC0613841