Learn More
Enzo Life Sciences LBP (mouse) monoclonal antibody (biG 33) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804502C100
Encompass_Preferred
Clone: biG 33. Isotype: IgG1. Immunogen: Recombinant mouse LBP (lipopolysaccharide binding protein) (in LBP knockout mouse). Species reactivity: Mouse. Applications: ELISA, WB, FUNC. Purity: Protein-G affinity purified. Formulation: Lyophilized. Contains PBS.
Catalog No. 50-200-9930