Learn More
Enzo Life Sciences LBP, soluble (mouse) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850305KI01
Encompass_Preferred
The LBP, soluble (mouse) ELISA kit is a complete assay for the quantitative determination of soluble as well as bound LBP levels. This kit is to be used with serum, plasma and cell culture medium from mouse or rat origins. Alternative name: Lipopolysaccharide binding protein. Assay Time: 2 hours + 12 to 15 minutes. Applications: ELISA . Species reactivity: Mouse, Rat .
Catalog No. 50-201-0157