Learn More
Enzo Life Sciences LBP, soluble ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850304KI01
Encompass_Preferred
The LBP, soluble ELISA kit is a complete assay for the quantitative determination of soluble LBP levels. This kit is to be used with serum, plasma and cell culture medium from human, bovine, dog, horse, porcine or rabbit origins. Alternative name: Lipopolysaccharide binding protein. Assay Time: 2 hours + 10 to 12 minutes. Applications: ELISA . Species reactivity: Human Rabbit, Porcine, Horse, Dog, Bovine.
Catalog No. 50-201-0156