Learn More
Hycult Biotech Inc LBP Various Species ELISA Kit, 1 x 96 detections, Sandwich ELISA, Assay time 3.5 hours, Standard range 1.6 to 100 ng/mL, Sensitivity 1.6 ng/mL

Supplier: Hycult Biotech Inc HK503
The HYCULT BIOTECH INC HK503 LBP ELISA kit is a ready-to-use, solid-phase enzyme-linked immunosorbent assay designed for quantitative determination of Lipopolysaccharide Binding Protein (LBP) in various biological samples. It offers a robust and efficient measurement solution for immunology and inflammation research across multiple species.
- Product type: ELISA kit
- Target analyte: Lipopolysaccharide Binding Protein (LBP)
- Assay principle: Sandwich ELISA
- Kit format: 1 x 96 detections
- Sample types: Cell culture supernatants, plasma, serum
- Assay time: 3.5 hours
- Standard range: 1.6 to 100 ng/mL
- Sensitivity: 1.6 ng/mL
- Cross-reactivity: Bovine, canine, human, rabbit, rat, and others
- Storage conditions: 4 °C
- Stability: 6 months
- Research use only: true
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.