Learn More
Echelon Biosciences Research Labs LBPA BEADS 1 mL
SDP
Catalog No. 50720696
Encompass
LBPA Beads are composed of agarose with 10 nanomoles of bound Lysobisphosphatidic Acid (LBPA) per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-696 Supplier Echelon Biosciences Research Labs Supplier No. PBLBPA1ML
LBPA Beads are composed of agarose with 10 nanomoles of bound Lysobisphosphatidic Acid (LBPA) per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable