Learn More
Oz Biosciences LentiBlast Premium Transduction Enhancer | 500 µL
SDP
Supplier:  Oz Biosciences LBPX500
Encompass
LentiBlast Premium is a novel patented chemical composition that dramatically increases lentiviral infection and transduction efficiency in any type of cells, adherent or in suspension, primary or cell lines. The properties of this reagent allow simultaneously neutralizing electrostatic repulsions between membrane and viral particles and enhancing viral fusion with cell membrane. Due to a favorable “membrane permeable effect” limiting the transmembrane potential changes, LentiBlast Premium is non-toxic and totally compatible with cell viability.
Catalog No. NC1736525