Learn More
Aviva Systems Biology Corp LBR Antibody - middle region (ARP45263_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45263_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LBR. Alternate names: DHCR14B, LMN2R, MGC9041, PHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62952