Learn More
Aviva Systems Biology Corp LBX1 Antibody - N-terminal region (ARP38817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LBX1. Alternate names: HPX-6, HPX6, LBX1H, homeoboxApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-760