Learn More
Aviva Systems Biology Corp LBX1 Antibody - middle region (ARP32808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LBX1. Alternate names: HPX-6, HPX6, LBX1H, homeoboxApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54775