Learn More
Aviva Systems Biology Corp Lbxcor1 Antibody - middle region (ARP37517_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37517_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lbxcor1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Lbxcor1. Alternate names: 4931440A04, AV273001, C230094B15Rik, Corl1, Lbxcor1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-647