Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Catalog No. p-4587714 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Application):
Food, Bev, Cosmetics, Pharm
Catalog No. For Use With (Application)
09-053-671 Food, Bev, Cosmetics, Pharm
1 options
Catalog No. 09-053-671 Supplier Lovibond™ Supplier No. 401100
Only null left
Add to Cart
Add to Cart
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
  • Each measurement combines 27 images illuminated with different colours and from different directions
  • Enables preview of measurement area and storage and recall of image after measurement
  • Measures and stores up to 20 reference standards with each date and time stamped with option to name
  • Stores up to 350 sample measurements on device and compares to reference standards
  • Samples can be tagged via on-screen keyboard for text or via built in voice recorder
  • For indoor use only
LC 100 and SV 100 Integrated Kit
  • Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
  • Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifications
Display TFT
For Use With (Application) Food, Bev, Cosmetics, Pharm
Includes Spectrophotometer, holder, sample cells, USB cord, carrying case
Measurement Time 1.8 sec.