Learn More
Alkali Scientific LC-8 3500 C3100 Centrifuge wit
SDP
Catalog No. 503029636
Encompass_Preferred
Alkalis LC-8 3500 C3100 of lab centrifuge is uniquely designed to meet a wide range of applications in both the clinical and research laboratory An eight position rotor is included and designed to directly accept 10ml and 15ml test-tubes (conical or round bottom) while optional adapters are also available for use with all popular sizes of blood collection and culture tubes Unlike most compact centrifuges in this class the rotor can be easily removed from the chamber for cleaning The back-lit liquid crystal display provides full control over the set run time and rotational speed The rotational speed can be set/viewed in rpm or rcf and acceleration/deceleration levels can be easily adjusted in the advanced settings mode To ensure safety the digital microprocessor ensures that the lid has been closed and that the safety lid lock is engaged prior to starting a centrifugation cycle In addition an advanced imbalance detection system constantly monitors the balance and vibration automat
Catalog No. 50-302-9636 Supplier Alkali Scientific Supplier No. RS7701
Alkalis LC-8 3500 C3100 of lab centrifuge is uniquely designed to meet a wide range of applications in both the clinical and research laboratory An eight position rotor is included and designed to directly accept 10ml and 15ml test-tubes (conical or round bottom) while optional adapters are also available for use with all popular sizes of blood collection and culture tubes Unlike most compact centrifuges in this class the rotor can be easily removed from the chamber for cleaning The back-lit liquid crystal display provides full control over the set run time and rotational speed The rotational speed can be set/viewed in rpm or rcf and acceleration/deceleration levels can be easily adjusted in the advanced settings mode To ensure safety the digital microprocessor ensures that the lid has been closed and that the safety lid lock is engaged prior to starting a centrifugation cycle In addition an advanced imbalance detection system constantly monitors the balance and vibration automat