Learn More
Stellar Scientific Benchmark Scientific Clinical Lab Centrifuge LC-8 (max speed 3500 rpm) for 8 x 15mL tubes
SDP
Catalog No. NC1379579
Encompass_Preferred
This clinical/research centrifuge features a generous 8 place rotor to increase capacity and a digital interface that automatically converts rpm to rcf. For added security, the lid-lock system will only open once the rotor has stopped and imbalance detection automatically shuts off the unit. The entire rotor can be easily removed for quick clean-upThe Benchmark LC-8 will hold 15ml conical and round bottom tubes, and may be equipped with optional adapter tubes to accommodate 5ml/7ml blood/culture or vacutainer tubes. Other DetailsSpeed:400 to 3500 rpm (1500xg) Accel. / Decel.:9 sec. / 25 sec. Speed Increment:100rpm increments Timer:20 sec. to 99 min. or continuous Dimensions: (w x d x h):11.6 x 14.4 x 8.3 in. 29.5 x 36.5 x 21 cm
Catalog No. NC1379579 Supplier Stellar Scientific Supplier No. BSCCF8
This clinical/research centrifuge features a generous 8 place rotor to increase capacity and a digital interface that automatically converts rpm to rcf. For added security, the lid-lock system will only open once the rotor has stopped and imbalance detection automatically shuts off the unit. The entire rotor can be easily removed for quick clean-upThe Benchmark LC-8 will hold 15ml conical and round bottom tubes, and may be equipped with optional adapter tubes to accommodate 5ml/7ml blood/culture or vacutainer tubes. Other DetailsSpeed:400 to 3500 rpm (1500xg) Accel. / Decel.:9 sec. / 25 sec. Speed Increment:100rpm increments Timer:20 sec. to 99 min. or continuous Dimensions: (w x d x h):11.6 x 14.4 x 8.3 in. 29.5 x 36.5 x 21 cm