Learn More
Roche Diagnostics LightCycler 8-Tube Strips (white), 120 x 8 tube strips and caps
Catalog No. 50588382 Shop All Roche Diagnostics Products
Encompass
LightCycler 8-Tube Strips are white polypropylene reaction devices linked to form a strip of 8 in a row. Their dimensions and material achieve optimized assay conditions using LightCycler 96 Instrument reaction volumes of 10 to 50 ul (recommended volume 20 ul) or using the LightCycler 480 Instrument. The LightCycler 8- Tube Strips are provided with strips of flat clear polypropylene caps.
Catalog No. 50-588-382 Supplier Roche Diagnostics Supplier No. 06612601001
LightCycler 8-Tube Strips are white polypropylene reaction devices linked to form a strip of 8 in a row. Their dimensions and material achieve optimized assay conditions using LightCycler 96 Instrument reaction volumes of 10 to 50 ul (recommended volume 20 ul) or using the LightCycler 480 Instrument. The LightCycler 8- Tube Strips are provided with strips of flat clear polypropylene caps.