Learn More
Bioanalytical Systems Inc LC AUX ELEC-DS CF-STYLE
SDP
Catalog No. 50854533
Encompass_Preferred
Electrode, Auxiliary; BASi; For Thin-Layer Flowcell; DS Cross-flow Style *Does not ship to Puerto Rico
Catalog No. 50-854-533 Supplier Bioanalytical Systems Inc Supplier No. MF1093
Electrode, Auxiliary; BASi; For Thin-Layer Flowcell; DS Cross-flow Style *Does not ship to Puerto Rico