Learn More
QA Bio Inc LudgerSep uR2 2.1x50mm UHPLC Column for Glycan Analysis
Supplier:  QA Bio Inc LSUR22.1X50
Encompass
uR2 2.1x50mm UHPLC Column for Glycan Analysis • The LudgerSep uR2 2.1x50mm column has been developed for UHPLC analysis of sialic acids labeled with DMB using the LudgerTag DMB Sialic Acid Labeling Kit. • Dimensions • 2.1mm x 50mm
Catalog No. 50-355-845