Learn More
Optimize Technologies EXP Titanium Hybrid Ferrule, 10pk
SDP
Catalog No. 507533778
Encompass_Preferred
The EXP™ Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between 1/16” tubing and any 10-32 port. EXP™ Fittings are simple and versatile. The patented Titanium Hybrid Ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The Hybrid Ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.
Catalog No. 50-753-3778 Supplier Optimize Technologies Supplier No. 152403832
The EXP™ Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between 1/16” tubing and any 10-32 port. EXP™ Fittings are simple and versatile. The patented Titanium Hybrid Ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The Hybrid Ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.