Learn More
Waters Corp WATERS CORPORATION LC Connections Kit, Waters Fraction Collector III, HPLC
Catalog No. 50466573 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed to facilitate the connection of a Waters Fraction Collector III to a Liquid Chromatography (LC) System.
  • Contains all necessary tubing, nuts, and ferrules required to connect a Waters Fraction Collector III to an LC system.
  • Designed for use with Waters Fraction Collector III and High Performance Liquid Chromatography (HPLC) systems.
  • Classified as a spare parts kit.
  • One unit per package.
  • Replacement as required.
Catalog No. 50-466-573 Supplier Waters Corp Supplier No. 700002476
This kit is designed to facilitate the connection of a Waters Fraction Collector III to a Liquid Chromatography (LC) System.
  • Contains all necessary tubing, nuts, and ferrules required to connect a Waters Fraction Collector III to an LC system.
  • Designed for use with Waters Fraction Collector III and High Performance Liquid Chromatography (HPLC) systems.
  • Classified as a spare parts kit.
  • One unit per package.
  • Replacement as required.