Learn More
Optimize Technologies EXP Discovery Kit - Contains 1 holder and 3 trap cartridge packs (total of 9 traps). Create your custom kit by choosing a holder (Direct Connect or In-Line) and any 3 of the cartridge configurations.
SDP
Catalog No. 507533522
Encompass_Preferred
The patent-pending Hand-Tight EXP™ Trap Column is rated for use at 20,000+ psi (1,400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with 1/16” stainless tubing for unparalleled convenience and efficiency. The EXP™ Cartridge System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet.
Catalog No. 50-753-3522 Supplier Optimize Technologies Supplier No. 150204042
Add to Cart
Add to Cart
The patent-pending Hand-Tight EXP™ Trap Column is rated for use at 20,000+ psi (1,400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with 1/16” stainless tubing for unparalleled convenience and efficiency. The EXP™ Cartridge System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet.