Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ LC Extension Cable

Catalog No. 01919415 Shop All OHAUS Products
Change view
Click to view available options
Length (English):
30 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
01-919-415 30 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-919-415 Supplier OHAUS™ Supplier No. 30424409
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nine meter long LC extension cable.

  • 9 meters (30 ft.)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MB Series, Discovery, Ranger, Defender DxxP
Connection Type Cable Extension 9m D52
Length (English) 30 ft.
Length (Metric) 9 m
Type Extension Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.