Learn More
Bioanalytical Systems Inc LC FLOW CELL-DUAL 3MM ELECT
SDP
Catalog No. 50854928
Encompass_Preferred
Kit, Flow Cell; Thin-layer; Dual 3mm electrode style; Includes includes glassy carbon electrode, gasket, reference electrodes, retaining hardware, polishing kit *Does not ship to Puerto Rico
Catalog No. 50-854-928 Supplier Bioanalytical Systems Inc Supplier No. MW5052
Kit, Flow Cell; Thin-layer; Dual 3mm electrode style; Includes includes glassy carbon electrode, gasket, reference electrodes, retaining hardware, polishing kit *Does not ship to Puerto Rico