Learn More
Bioanalytical Systems Inc LC GAS INLET BARB FITTING
SDP
Catalog No. 50854832
Encompass_Preferred
Fitting: Gas inlet barb; BASi; For use with voltammetry cells and RDE-2 *Does not ship to Puerto Rico
Catalog No. 50-854-832 Supplier Bioanalytical Systems Inc Supplier No. MR4404
Fitting: Gas inlet barb; BASi; For use with voltammetry cells and RDE-2 *Does not ship to Puerto Rico