Learn More
Optimize Technologies EXP Trap Cartridge, 4.6mm x 5mm, 3.0µm EXP, SCX, 3pk
SDP
Catalog No. 507533582
Encompass_Preferred
The Hand-Tight EXP™ Trap Column is rated to 20,000+ psi. This unique design connects directly to any injection valve (with 10-32 threads) or in-line with 1/16” tubing for unparalleled convenience and efficiency. The EXP™ System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. Free-Turn™ architecture allows users to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. The EXP™ Trap Column family now offers packed beds available in 2, 5, 10, 15, 20 and 30mm lengths with diameters ranging from 1-4.6mm and bed volumes from 1.6-600µL. Please contact your representative directly if you're interested in a length other than 5mm.
Catalog No. 50-753-3582 Supplier Optimize Technologies Supplier No. 1503983HM
The Hand-Tight EXP™ Trap Column is rated to 20,000+ psi. This unique design connects directly to any injection valve (with 10-32 threads) or in-line with 1/16” tubing for unparalleled convenience and efficiency. The EXP™ System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. Free-Turn™ architecture allows users to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. The EXP™ Trap Column family now offers packed beds available in 2, 5, 10, 15, 20 and 30mm lengths with diameters ranging from 1-4.6mm and bed volumes from 1.6-600µL. Please contact your representative directly if you're interested in a length other than 5mm.