Learn More
Enzo Life Sciences LC3-I (human) (recombinant) (His-tag) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAPR1000200
Encompass_Preferred
Alternative name: Microtubule-associated protein 1A/1B-light chain 3. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥95% (SDS-PAGE). MW: ~16kDa (predicted), 20kDa (observed). Formulation: Liquid. In 25mM TRIS, pH 7.5, containing 250mM sodium chloride, 1.0mM EDTA, and 1.0mM DTT. Source: Produced in E. coli. Human LC3-I is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7177