Learn More
Enzo Life Sciences LC3 monoclonal antibody (5H3) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX803082C100
Encompass_Preferred
Clone: 5H3. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to an internal sequence identical in LC3A, LC3B and LC3C conjugated to hemocyanin. Species reactivity: Human. Specificity: Recognizes both forms of endogenous LC3. Applications: ICC, IHC, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Liquid. In PBS containing 50% glycerol, 0.09% sodium azide, PEG and sucrose.
Catalog No. 50-200-9732